SY3371479286 Synthonix Corporation

2-Methyl-4-hydroxymethylpyridine

  • MDL number MFCD08062415
  • Cas Number : 105250-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/4019.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/4019.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart