SY3371479292 Synthonix Corporation

1-(Triisopropylsilyl)pyrrole-3-boronic acid

  • MDL number MFCD01114667
  • Cas Number : 138900-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/N210.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/N210.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart