SY3371479323 Synthonix Corporation

7-Oxo-2,6-diazaspiro[3.4]octane-2-carboxylic acid tert-butyl ester

  • MDL number MFCD18325190
  • Cas Number : 1234616-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6102.html

Safety Information

pcodes P280 ,
MSDS https://synthonix.com/6102.html
hstatements H301 , H319 ,
Availability details for
Quantity
  • Availability
  • Add to Cart