SY3371479344 Synthonix Corporation

cis-Octahydro-1H-isoindole

  • Cas Number : 1470-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6365.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6365.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart