SY3371479370 Synthonix Corporation

5-Methylthiazole-4-carboxylic acid ethyl ester

  • MDL number MFCD18427704
  • Cas Number : 61323-26-0
  • Molecular Weight : 171.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/4176.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/4176.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart