SY3390161401 Synthonix Corporation

2-Amino-4-methylthiophene Hydrochloride

  • MDL number MFCD21604144
  • Cas Number : 930299-88-0
  • Molecular Weight : 149.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0597.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/0597.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart