SY3390161405 Synthonix Corporation

6-(Aminomethyl)pyrimidin-4-amine dihydrochloride

  • MDL number MFCD22199240
  • Cas Number : 1523618-18-9
  • Molecular Weight : 160.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0583.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/0583.html
pictograms GHS06, GHS07, GHS08, GHS09
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart