SY3390161407 Synthonix Corporation

4-(Oxetan-3-yl)piperazine ditrifluoroacetic acid salt

  • MDL number MFCD22573363
  • Cas Number : 1404373-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6546.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6546.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart