SY3390161408 Synthonix Corporation

(S)-3-Amino-1-methyl-piperidine dihydrochloride

  • MDL number MFCD22199150
  • Cas Number : 1157849-51-8
  • Molecular Weight : 150.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/1654.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/1654.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart