SY3390161416 Synthonix Corporation

2-Amino-5-bromo-4-chloro-3-nitropyridine

  • MDL number MFCD11110693
  • Cas Number : 942947-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7610.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7610.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart