SY3390161431 Synthonix Corporation

1-Boc-3-(aminomethyl)-3-methylazetidine

  • MDL number MFCD12408560
  • Cas Number : 1158758-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6945.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6945.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart