SY3390161457 Synthonix Corporation

(S)-3-(Boc-amino)piperidine hydrochloride

  • MDL number MFCD08275944
  • Cas Number : 1416450-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/7247.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7247.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart