SY3390161472 Synthonix Corporation

5-Bromoimidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD11226918
  • Cas Number : 878197-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7536.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7536.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart