SY3390161484 Synthonix Corporation

5-Bromo-1-methyl-3-nitropyridin-2(1H)-one

  • MDL number MFCD09835081
  • Cas Number : 153888-45-0
  • Molecular Weight : 233.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7465.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7465.html
pictograms GHS05, GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart