SY3390161504 Synthonix Corporation

5-Chloro-3-methyl-4-nitro-1H-pyrazole

  • MDL number MFCD00462202
  • Cas Number : 6814-58-0
  • Molecular Weight : 161.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7549.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7549.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart