SY3390161527 Synthonix Corporation

Cyclopropanecarboxylic acid hydrazide

  • MDL number MFCD00045392
  • Cas Number : 6952-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7568.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7568.html
hstatements H315 , H317 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart