SY3390161572 Synthonix Corporation

4-Hydroxy-4-methylpiperidine hydrochloride

  • MDL number MFCD11044137
  • Cas Number : 586375-35-1
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7439.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/7439.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart