SY3390161588 Synthonix Corporation

4-Iodo-2,7-naphthyridin-1(2H)-one

  • MDL number MFCD17015944
  • Cas Number : 959558-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/7562.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7562.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart