SY3390161589 Synthonix Corporation

2-Iodo-1-(phenylsulfonyl)-7-azaindole

  • MDL number MFCD11559008
  • Cas Number : 282734-63-8
  • Molecular Weight : 384.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7533.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/7533.html
pictograms GHS07
hstatements H301, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart