SY3390161606 Synthonix Corporation

1-Cbz-3-methylazetidine-3-carboxylic acid

  • MDL number MFCD17168405
  • Cas Number : 1143525-35-2
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7521.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7521.html
pictograms GHS07
hstatements H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart