SY3390161620 Synthonix Corporation

7-Methyl-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD05723324
  • Cas Number : 27582-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/7620.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7620.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart