SY3390161645 Synthonix Corporation

7-Methyl-7H-pyrrolo[2,3-d]pyrimidine-4-carboxylic acid

  • MDL number MFCD17016002
  • Cas Number : 1095822-19-7
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7616.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7616.html
pictograms GHS06, GHS07, GHS08, GHS09
hstatements H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart