SY3390161659 Synthonix Corporation

Potassium bis(fluorosulfonyl)imide

  • MDL number MFCD17018950
  • Cas Number : 14984-76-0
  • Molecular Weight : 219.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7137.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7137.html
pictograms GHS05, GHS07
hstatements H301, H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart