SY3390161666 Synthonix Corporation

2-Pyrimidineacetonitrile

  • MDL number MFCD09759105
  • Cas Number : 59566-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7449.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7449.html
hstatements H302 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart