SY3390161680 Synthonix Corporation

(R)-(1-Tetrahydrofuran-3-yl)methanamine hydrochloride

  • MDL number MFCD20526349
  • Cas Number : 1400744-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7262.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7262.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart