SY3390161694 Synthonix Corporation

1-{[(2-Trimethylsilyl)ethoxy]methyl}-1H-pyrrolo[2,3-b]pyridin-2-one

  • MDL number MFCD17214392
  • Cas Number : 879132-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7635.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/7635.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart