SY3390161700 Synthonix Corporation

1-Benzyl-1H-pyrazole-4-carbonyl chloride

  • MDL number MFCD12403335
  • Cas Number : 1174665-95-2
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8224.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/8224.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart