SY3390161704 Synthonix Corporation

cis-4-Amino-1-Boc-3-methoxypiperidine

  • MDL number MFCD22415234
  • Cas Number : 1171124-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8099.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8099.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart