SY3390161722 Synthonix Corporation

(3S,4S)-3-Amino-4-hydroxy-tetrahydropyran

  • MDL number MFCD19215826
  • Cas Number : 1240390-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7869.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/7869.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart