SY3390161727 Synthonix Corporation

exo-3-Amino-9-methyl-9-azabicyclo[3.3.1]nonane

  • MDL number MFCD12198832
  • Cas Number : 76272-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8192.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8192.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart