SY3390161735 Synthonix Corporation

3-Aminomethyl-2-methoxypyridine dihydrochloride

  • MDL number MFCD15173358
  • Cas Number : 1158447-85-8
  • Molecular Weight : 211.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7689.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7689.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart