SY3390161744 Synthonix Corporation

7-Azabicyclo[2.2.1]heptane hydrochloride

  • MDL number MFCD00154183
  • Cas Number : 27514-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8159.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8159.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart