SY3390161751 Synthonix Corporation

1-(Boc-amino)-3,3-difluorocyclobutanecarboxylic acid

  • MDL number MFCD22415254
  • Cas Number : 1363380-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8011.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8011.html
hstatements H302 , H315 , H317 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart