SY3390161757 Synthonix Corporation

3-Boc-3,7-diazabicyclo[4.2.0]octane

  • MDL number MFCD08234914
  • Cas Number : 885271-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/7992.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7992.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart