SY3390161769 Synthonix Corporation

1-Boc-octahydro-pyrrolo[3,2-b]pyridine

  • MDL number MFCD18382710
  • Cas Number : 1251010-63-5
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7883.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7883.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart