SY3390161774 Synthonix Corporation

2-Boc-1,2,3,4-tetrahydro-isoquinoline-7-carboxylic acid

  • MDL number MFCD03094736
  • Cas Number : 149353-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/7889.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7889.html
pictograms GHS07
hstatements H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart