SY3390161792 Synthonix Corporation

5-Bromo-2-methyl-4-nitropyridine-N-oxide

  • MDL number MFCD11977426
  • Cas Number : 62516-08-9
  • Molecular Weight : 233.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8178.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/8178.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart