SY3390161824 Synthonix Corporation

4-Chloro-2-(methylthio)pyrrolo[1,2-f][1,2,4]triazine

  • MDL number MFCD11706978
  • Cas Number : 1120214-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8054.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8054.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart