SY3390161864 Synthonix Corporation

4,4-Difluoro-L-proline methyl ester trifluoroacetate

  • MDL number MFCD22415243
  • Cas Number : 915230-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8138.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8138.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart