SY3390161870 Synthonix Corporation

1,3-Dimethyl-6-amino-1H-indazole

  • MDL number MFCD18432764
  • Cas Number : 221681-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8180.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8180.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart