SY3390161871 Synthonix Corporation

6,6-Dimethyl-3-azabicyclo[3.1.0]hexane

  • MDL number MFCD13176114
  • Cas Number : 943516-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/7977.html

Safety Information

MSDS https://synthonix.com/7977.html
Availability details for
Quantity
  • Availability
  • Add to Cart