SY3390161901 Synthonix Corporation

exo-8-Boc-8-azabicyclo[3.2.1]octane-3-carboxylic acid methyl ester

  • Cas Number : 1548292-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8094.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/8094.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart