SY3390161914 Synthonix Corporation

3-(Hydroxymethyl)-1H-indazole

  • MDL number MFCD05260509
  • Cas Number : 64132-13-4
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7713.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/7713.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart