SY3390161920 Synthonix Corporation

3-Hyroxymethyl-1H-pyrazole

  • MDL number MFCD08436148
  • Cas Number : 23585-49-1
  • Molecular Weight : 98.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7696.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/7696.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart