SY3390161928 Synthonix Corporation

2-Iodo-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD16875827
  • Cas Number : 1227270-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8108.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8108.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart