SY3390161948 Synthonix Corporation

(1S,4S)-5-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide

  • MDL number MFCD01863510
  • Cas Number : 125224-62-6
  • Molecular Weight : 274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7868.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7868.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart