SY3390161953 Synthonix Corporation

(2S,4S)-1-Boc-2-methyl-4-hydroxypiperidine

  • MDL number MFCD22415236
  • Cas Number : 790667-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8129.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/8129.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart