SY3390161973 Synthonix Corporation

2-[(E)-2-Nitrovinyl]thiophene

  • MDL number MFCD00069434
  • Cas Number : 34312-77-1
  • Molecular Weight : 155.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7715.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/7715.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart