SY3390162063 Synthonix Corporation

5-Hydroxymethyl-2-BOC-hexahydrocyclopenta[C]pyrrole

  • MDL number MFCD20926145
  • Cas Number : 1365570-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7776.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/7776.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart