SY3390162070 Synthonix Corporation

2-Oxabicyclo[2.2.1]heptane-5-carboxylic acid, 3-oxo-, (1R,4R,5R)- cinchonidine salt

  • MDL number No MDL found
  • Cas Number : 1233219-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7821.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7821.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart